ENA238710086 Enamine

4-[(2E)-2-[(3-fluorophenyl)methylidene]hydrazin-1-yl]benzoic acid

  • MDL number MFCD02696391
  • Cas Number : 325729-60-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 229...231
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-41961

Safety Information

msds https://www.enaminestore.com/catalog/EN300-41961
Availability details for
Quantity
  • Availability
  • Add to Cart