ENA238710096 Enamine

N-benzyl-4-(2-sulfanyl-1H-imidazol-1-yl)benzamide

  • MDL number MFCD23147878
  • Cas Number : 1146289-95-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-42847

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-42847
Availability details for
Quantity
  • Availability
  • Add to Cart