ENA238710101 Enamine

N-[(4-fluorophenyl)methyl]-3-(2-sulfanyl-1H-imidazol-1-yl)benzamide

  • MDL number MFCD11857977
  • Cas Number : 1146290-01-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-42867

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-42867
Availability details for
Quantity
  • Availability
  • Add to Cart