ENA238710102 Enamine

N-[(2-fluorophenyl)methyl]-3-(2-sulfanyl-1H-imidazol-1-yl)benzamide

  • MDL number MFCD11857978
  • Cas Number : 1146290-02-9
  • Molecular Weight : 327.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-42869

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-42869
Availability details for
Quantity
  • Availability
  • Add to Cart