ENA238710106 Enamine

N-(2-aminophenyl)-1-(2,2-dichlorocyclopropyl)methanesulfonamide

  • MDL number MFCD11857980
  • Cas Number : 1146290-04-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-42875

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-42875
Availability details for
Quantity
  • Availability
  • Add to Cart