ENA238710110 Enamine

(2,2-dichlorocyclopropyl)methanesulfonyl chloride

  • MDL number MFCD11857994
  • Cas Number : 1146290-10-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-42967

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-42967
Availability details for
Quantity
  • Availability
  • Add to Cart