ENA238710112 Enamine

2-{[(2,2-dichlorocyclopropyl)methyl]sulfanyl}aniline

  • MDL number MFCD11857996
  • Cas Number : 1146290-11-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-42969

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-42969
Availability details for
Quantity
  • Availability
  • Add to Cart