ENA238710121 Enamine

(dodecylsulfanyl)-N-phenylmethanimidamide hydrobromide

  • MDL number MFCD11858029
  • Cas Number : 1173621-67-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 65...67
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-43183

Safety Information

msds https://www.enaminestore.com/catalog/EN300-43183
Availability details for
Quantity
  • Availability
  • Add to Cart