ENA238710145 Enamine

2-chloro-N-[(2Z)-3-(1-naphthylmethyl)-1,3-thiazol-2(3H)-ylidene]acetamide

  • MDL number MFCD06655241
  • Cas Number : 847781-98-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-11491

Safety Information

msds https://www.enaminestore.com/catalog/EN300-11491
Availability details for
Quantity
  • Availability
  • Add to Cart