ENA238710160 Enamine

N-cyclopropylthiophene-2-carboximidoyl chloride

  • MDL number MFCD07366398
  • Cas Number : 885524-73-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 225...227
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-14695

Safety Information

msds https://www.enaminestore.com/catalog/EN300-14695
Availability details for
Quantity
  • Availability
  • Add to Cart