ENA238710161 Enamine

4-fluoro-N-methyl-N'-[1-(propan-2-yl)-1H-pyrazol-5-yl]benzene-1-carboximidamide

  • Cas Number : 1006463-55-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-14753

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-14753
Availability details for
Quantity
  • Availability
  • Add to Cart