ENA238710163 Enamine

N-methyl-N'-(3-methyl-1-phenyl-1H-pyrazol-5-yl)benzenecarboximidamide

  • Cas Number : 380416-23-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-14755

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-14755
Availability details for
Quantity
  • Availability
  • Add to Cart