ENA238710164 Enamine

4-fluoro-N-methyl-N'-(3-methyl-1-phenyl-1H-pyrazol-5-yl)benzene-1-carboximidamide

  • Cas Number : 380394-41-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-14756

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-14756
Availability details for
Quantity
  • Availability
  • Add to Cart