ENA238710172 Enamine

N-(2-hydroxyethyl)-3-(thiophen-2-yl)prop-2-enamide

  • Cas Number : 327090-55-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-15592

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-15592
Availability details for
Quantity
  • Availability
  • Add to Cart