ENA238710180 Enamine

3-chloro-N-(3-methoxypropyl)benzene-1-sulfonamide

  • MDL number MFCD06357336
  • Cas Number : 790263-28-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-15822

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-15822
Availability details for
Quantity
  • Availability
  • Add to Cart