ENA238710187 Enamine

2-cyano-N-phenylbenzene-1-sulfonamide

  • MDL number MFCD04940987
  • Cas Number : 69360-16-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 248...250
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-15978

Safety Information

msds https://www.enaminestore.com/catalog/EN300-15978
Availability details for
Quantity
  • Availability
  • Add to Cart