ENA238710188 Enamine

N-(4-methoxyphenyl)thiophene-2-sulfonamide

  • MDL number MFCD01159690
  • Cas Number : 39810-50-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 120-121
Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-16004

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-16004
Availability details for
Quantity
  • Availability
  • Add to Cart