ENA238710191 Enamine

3-[(4-fluorophenyl)sulfanyl]propanenitrile

  • MDL number MFCD02755222
  • Cas Number : 151922-79-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-16190

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-16190
Availability details for
Quantity
  • Availability
  • Add to Cart