ENA238710192 Enamine

6-(3-bromophenyl)-1,3-diazinane-2,4-dione

  • MDL number MFCD05262779
  • Cas Number : 743440-60-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-16470

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-16470
Availability details for
Quantity
  • Availability
  • Add to Cart