ENA238710195 Enamine

(2E)-3-(naphthalen-2-ylcarbamoyl)prop-2-enoic acid

  • MDL number MFCD00627801
  • Cas Number : 292870-59-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-16564

Safety Information

msds https://www.enaminestore.com/catalog/EN300-16564
Availability details for
Quantity
  • Availability
  • Add to Cart