ENA238710200 Enamine

N-cyclopropyl-N'-(3-methyl-1-phenyl-1H-pyrazol-5-yl)benzenecarboximidamide

  • MDL number MFCD02709346
  • Cas Number : 380420-21-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-16852

Safety Information

msds https://www.enaminestore.com/catalog/EN300-16852
Availability details for
Quantity
  • Availability
  • Add to Cart