ENA238710206 Enamine

3-bromo-5-ethoxy-4-hydroxybenzaldehyde

  • MDL number MFCD00173904
  • Cas Number : 3111-37-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 141...143
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-17133

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-17133
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart