ENA238710222 Enamine

2-cyano-2-cyclohexylideneethanethioamide

  • MDL number MFCD00569790
  • Cas Number : 129224-53-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-17820

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-17820
Availability details for
Quantity
  • Availability
  • Add to Cart