ENA238710224 Enamine

N-(3-amino-4-chlorophenyl)butanamide

  • MDL number MFCD03723773
  • Cas Number : 637316-74-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 96...98
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-18161

Safety Information

msds https://www.enaminestore.com/catalog/EN300-18161
Availability details for
Quantity
  • Availability
  • Add to Cart