ENA238710234 Enamine

2-chloro-1-[1-(2,3-dihydro-1H-inden-5-yl)-2,5-dimethyl-1H-pyrrol-3-yl]ethan-1-one

  • MDL number MFCD06680114
  • Cas Number : 852840-44-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-18699

Safety Information

msds https://www.enaminestore.com/catalog/EN300-18699
Availability details for
Quantity
  • Availability
  • Add to Cart