ENA238710259 Enamine

4-(acetyloxy)-1-(4-methylbenzenesulfonyl)pyrrolidine-2-carboxylic acid

  • MDL number MFCD07685839
  • Cas Number : 947279-97-2
  • Molecular Weight : 327.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-24013

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-24013
Availability details for
Quantity
  • Availability
  • Add to Cart