ENA238710266 Enamine

N-methyl-N'-(3-methyl-1,2-oxazol-5-yl)benzenecarboximidamide

  • Cas Number : 878465-57-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-24296

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-24296
Availability details for
Quantity
  • Availability
  • Add to Cart