ENA238710283 Enamine

phenylmethanesulfonamide

  • MDL number MFCD00041893
  • Cas Number : 4563-33-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 104...106
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-26996

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-26996
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart