ENA238710291 Enamine

[(2-methoxyphenyl)methylidene](methyl)amine

  • MDL number MFCD09373420
  • Cas Number : 1268854-18-7
  • Molecular Weight : 149.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-27442

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-27442
Availability details for
Quantity
  • Availability
  • Add to Cart