ENA238710307 Enamine

3-[(prop-2-yn-1-yl)amino]benzoic acid

  • MDL number MFCD08445209
  • Cas Number : 923112-68-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-28864

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-28864
Availability details for
Quantity
  • Availability
  • Add to Cart