ENA238710311 Enamine

2-(chloromethyl)-4-methylquinazoline

  • MDL number MFCD09807547
  • Cas Number : 109113-72-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 60...62
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-29653

Safety Information

msds https://www.enaminestore.com/catalog/EN300-29653
Availability details for
Quantity
  • Availability
  • Add to Cart