ENA238710317 Enamine

ethyl 5-amino-3-methylthiophene-2-carboxylate hydrochloride

  • MDL number MFCD03425862
  • Cas Number : 41940-43-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 55-56
Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-30086

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-30086
Availability details for
Quantity
  • Availability
  • Add to Cart