ENA238710323 Enamine

(4-methylphenyl)(phenyl)methanamine hydrochloride

  • MDL number MFCD00182941
  • Cas Number : 5267-49-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 80-81
Purity 95
Salt Data HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-30201

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-30201
Availability details for
Quantity
  • Availability
  • Add to Cart