ENA238710333 Enamine

2-(chloromethyl)-1-(difluoromethyl)-1H-imidazole hydrochloride

  • MDL number MFCD09284930
  • Cas Number : 1049718-73-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-30343

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-30343
Availability details for
Quantity
  • Availability
  • Add to Cart