ENA238710346 Enamine

N-(4-cyanophenyl)propanamide

  • MDL number MFCD01812189
  • Cas Number : 314764-76-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 174...176
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-31324

Safety Information

msds https://www.enaminestore.com/catalog/EN300-31324
Availability details for
Quantity
  • Availability
  • Add to Cart