ENA238710348 Enamine

2-(2lambda4,1,3-benzothiadiazole-4-sulfonamido)propanoic acid

  • MDL number MFCD02724895
  • Cas Number : 1396966-20-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-00606

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-00606
Availability details for
Quantity
  • Availability
  • Add to Cart