ENA238710352 Enamine

[(3-fluorophenyl)methyl](propan-2-yl)amine

  • MDL number MFCD08060644
  • Cas Number : 90389-87-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-32096

Safety Information

signalword Danger
msds https://www.enaminestore.com/catalog/EN300-32096
pictograms GHS05,GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart