ENA238710354 Enamine

6-methoxy-1,2,3,4-tetrahydronaphthalen-1-amine

  • MDL number MFCD07778594
  • Cas Number : 52373-02-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-32144

Safety Information

msds https://www.enaminestore.com/catalog/EN300-32144
Availability details for
Quantity
  • Availability
  • Add to Cart