ENA238710362 Enamine

4-chloro-N-(furan-2-ylmethyl)aniline

  • MDL number MFCD04563642
  • Cas Number : 33829-87-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-32899

Safety Information

msds https://www.enaminestore.com/catalog/EN300-32899
Availability details for
Quantity
  • Availability
  • Add to Cart