ENA238710365 Enamine

4-methyl-N-(thiophen-2-ylmethyl)aniline

  • MDL number MFCD08690688
  • Cas Number : 3139-30-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-33043

Safety Information

msds https://www.enaminestore.com/catalog/EN300-33043
Availability details for
Quantity
  • Availability
  • Add to Cart