ENA238710368 Enamine

4-[(cyclopropylamino)methyl]-N,N-dimethylaniline

  • MDL number MFCD00465458
  • Cas Number : 892593-62-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-33124

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-33124
Availability details for
Quantity
  • Availability
  • Add to Cart