ENA238710384 Enamine

[1-(2,4-dimethylphenyl)ethyl](methyl)amine

  • MDL number MFCD09049453
  • Cas Number : 926253-30-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-33238

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-33238
Availability details for
Quantity
  • Availability
  • Add to Cart