ENA238710392 Enamine

methyl 2-amino-4-(difluoromethoxy)-5-methoxybenzoate

  • MDL number MFCD08444622
  • Cas Number : 923683-37-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-33490

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-33490
Availability details for
Quantity
  • Availability
  • Add to Cart