ENA238710408 Enamine

1-(morpholin-4-yl)-1-phenylpropan-2-amine

  • MDL number MFCD06654908
  • Cas Number : 100800-07-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 130-131
Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-34333

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-34333
Availability details for
Quantity
  • Availability
  • Add to Cart