ENA238710414 Enamine

5-(aminomethyl)thiophene-2-sulfonamide hydrochloride

  • MDL number MFCD08262010
  • Cas Number : 21151-36-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 120-121
Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-34531

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-34531
Availability details for
Quantity
  • Availability
  • Add to Cart