ENA238710426 Enamine

2-{5H,6H,7H,8H,9H-[1,2,4]triazolo[4,3-a]azepin-3-yl}aniline

  • MDL number MFCD02251853
  • Cas Number : 303019-48-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-34644

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-34644
Availability details for
Quantity
  • Availability
  • Add to Cart