ENA238710438 Enamine

3-chloro-4-(trifluoromethyl)-5H,6H,7H-cyclopenta[c]pyridazine

  • MDL number MFCD09040416
  • Cas Number : 939758-08-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-34889

Safety Information

msds https://www.enaminestore.com/catalog/EN300-34889
Availability details for
Quantity
  • Availability
  • Add to Cart