ENA238710440 Enamine

2-benzoyl-1-benzofuran-3-amine

  • MDL number MFCD00445250
  • Cas Number : 49615-93-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-34893

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-34893
Availability details for
Quantity
  • Availability
  • Add to Cart