ENA238710454 Enamine

2-(hydroxymethyl)-7-methyl-4H-pyrido[1,2-a]pyrimidin-4-one

  • MDL number MFCD08442772
  • Cas Number : 926185-92-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. Normal

Safety Information

msds https://www.enaminestore.com/catalog/EN300-35222
Availability details for
Quantity
  • Availability
  • Add to Cart