ENA238710478 Enamine

N-[(2Z)-3-cyclopentyl-4-methyl-1,3-thiazol-2(3H)-ylidene]-N-phenylamine

  • MDL number MFCD07286154
  • Cas Number : 454237-73-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 51...53
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-35568

Safety Information

msds https://www.enaminestore.com/catalog/EN300-35568
Availability details for
Quantity
  • Availability
  • Add to Cart